siRNA Details
| Gene Symbol: Smo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176996.4 | NP_795970.3 | 8 | 1911 | ||
| GenBank mRNA | AK081050.1 | - | 2 | 99 | ||
| AK193396.1 | - | 4 | 298 | |||
| AK194911.1 | - | 4 | 303 | |||
| BC096028.1 | - | 8 | 1430 | 6850065 | ||
| BQ947565.1 | - | 3 | 173 | |||
| CA749812.1 | - | 5 | 511 | |||
| FJ883552.1 | - | 8 | 1420 | |||
More Information
Set Membership: |
|

Smo
Genomic Map