siRNA Details
| Gene Symbol: Igf2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183029.2 | NP_898850.2 | 2, 3 | 585 | ||
| GenBank mRNA | AK044984.1 | - | 1, 2 | 101 | ||
| AK049196.1 | - | 2, 3 | 282 | |||
| AK077404.1 | - | 2, 3 | 264 | |||
| AK159654.1 | - | 2, 3 | 320 | |||
| AK170531.1 | - | 1, 2 | 89 | |||
| AY531659.1 | - | 2, 3 | 249 | |||
| BC023758.1 | - | 2, 3 | 283 | |||
| BC054552.1 | - | 2, 3 | 305 | |||
| CB290059.1 | - | 2, 3 | 585 | |||

Igf2bp2
Genomic Map