siRNA Details
Gene Symbol: Rc3h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100591.1 | NP_001094061.1 | 13 | 2417 | ||
NM_001290642.1 | NP_001277571.1 | 13 | 2417 | |||
GenBank mRNA | AK053071.1 | - | 13 | 2417 | ||
BC117951.1 | - | 13 | 2410 | 40098089 | ||
BC117952.1 | - | 13 | 2410 | 40098090 |
