siRNA Details
| Gene Symbol: 5031434C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087115.1 | - | 2 | 925 | ||
| AK165118.1 | - | 952 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_177012.1 | - | 925 | Chr. - - [-] on Build | ||

5031434C07Rik
Genomic Map