siRNA Details
| Gene Symbol: Slc17a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177016.3 | NP_795990.2 | 3 | 385 | ||
| GenBank mRNA | AK033669.1 | - | 3 | 388 | ||
| AK050272.1 | - | 3 | 384 | |||
| AK162320.1 | - | 3 | 384 | |||
| BC147415.1 | - | 3 | 346 | 9056491 | ||
| BC147416.1 | - | 3 | 346 | 9056498 | ||

Slc17a4
Genomic Map