siRNA Details
| Gene Symbol: Zcchc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138590.4 | NP_613056.2 | 4 | 835 | ||
| GenBank mRNA | AK029808.1 | - | 4 | 842 | ||
| AK032353.1 | - | 4 | 807 | |||
| AK039158.1 | - | 4 | 756 | |||
| AK043238.1 | - | 4 | 748 | |||
| AK080785.1 | - | 5 | 924 | |||
| AK157728.1 | - | 4 | 738 | |||
| BC050200.1 | - | 3 | 710 | |||
| BC117020.1 | - | 4 | 738 | 40126339 | ||
| BC117022.1 | - | 4 | 738 | 40126341 | ||
| BC144774.1 | - | 4 | 742 | 9053322 | ||
| CF732773.1 | - | 3 | 426 | |||

Zcchc7
Genomic Map