siRNA Details
| Gene Symbol: Ism1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276489.1 | NP_001263418.1 | 4 | 1185 | ||
| GenBank mRNA | AK077398.1 | - | 4 | 1186 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ism1| Gene Symbol: Ism1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276489.1 | NP_001263418.1 | 4 | 1185 | ||
| GenBank mRNA | AK077398.1 | - | 4 | 1186 | ||