siRNA Details
| Gene Symbol: Flad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177041.3 | NP_796015.2 | 2, 3 | 314 | ||
| GenBank mRNA | AK044501.1 | - | 323 | |||
| AK161929.1 | - | 2, 3 | 358 | |||
| AK211873.1 | - | 1, 2 | 181 | |||
| BC025817.1 | - | 2, 3 | 267 | 5099127 | ||
| BC046769.1 | - | 2, 3 | 314 | 6809196 | ||

Flad1
Genomic Map