siRNA Details
| Gene Symbol: Ttll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178869.4 | NP_849200.2 | 9 | 1254 | ||
| GenBank mRNA | AK031917.1 | - | 9 | 1225 | ||
| AK047046.1 | - | 9 | 1239 | |||
| AK084005.1 | - | 9 | 1222 | |||
| AK140472.1 | - | 9 | 1222 | |||
| AK168786.1 | - | 9 | 1225 | |||
| AK169150.1 | - | 9 | 1241 | |||
| BC010510.1 | - | 9 | 1203 | 4191464 | ||

Ttll1
Genomic Map