siRNA Details
| Gene Symbol: Ttll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178869.4 | NP_849200.2 | 8 | 1079 | ||
| GenBank mRNA | AK031917.1 | - | 8 | 1050 | ||
| AK047046.1 | - | 8 | 1064 | |||
| AK084005.1 | - | 8 | 1047 | |||
| AK140472.1 | - | 8 | 1047 | |||
| AK168786.1 | - | 8 | 1050 | |||
| AK169150.1 | - | 8 | 1066 | |||
| BC010510.1 | - | 8 | 1028 | 4191464 | ||

Ttll1
Genomic Map