siRNA Details
| Gene Symbol: Casc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205369.1 | NP_001192298.1 | 1 | 295 | ||
| NM_001205370.1 | NP_001192299.1 | 1 | 295 | |||
| NM_177054.5 | NP_796028.2 | 1 | 295 | |||
| NM_199038.3 | NP_950239.2 | 1 | 295 | |||
| GenBank mRNA | AK051613.1 | - | 1 | 137 | ||
| AK080364.1 | - | 1 | 146 | |||
| AK160708.1 | - | 1 | 136 | |||
| AY484583.1 | - | 1 | 135 | |||
| AY484585.1 | - | 1 | 135 | |||
| BC034556.1 | - | 1 | 158 | |||
| BC064446.1 | - | 1 | 295 | 6843743 | ||

Casc4
Genomic Map