siRNA Details
| Gene Symbol: BC106179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041922.1 | - | 2 | 185 | ||
| AK161755.1 | - | 2 | 470 | |||
| BC106179.1 | - | 2 | 466 | 6509459 | ||
| Gene Symbol: B630019A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046764.1 | - | 3 | 2234 | ||

BC106179
Genomic Map