siRNA Details
| Gene Symbol: Rps6kc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178775.4 | NP_848890.3 | 6 | 646 | ||
| GenBank mRNA | AK030313.1 | - | 624 | |||
| AK044809.1 | - | 3 | 365 | |||
| AK149858.1 | - | 6 | 611 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rps6kc1| Gene Symbol: Rps6kc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178775.4 | NP_848890.3 | 6 | 646 | ||
| GenBank mRNA | AK030313.1 | - | 624 | |||
| AK044809.1 | - | 3 | 365 | |||
| AK149858.1 | - | 6 | 611 | |||