siRNA Details
| Gene Symbol: Rps6kc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178775.4 | NP_848890.3 | 11 | 2622 | ||
| GenBank mRNA | AK044809.1 | - | 8 | 2341 | ||
| AK049003.1 | - | 4 | 1570 | |||
| AK149858.1 | - | 11 | 2587 | |||
| BC058403.1 | - | 2 | 1321 | 6812243 | ||

Rps6kc1
Genomic Map