siRNA Details
| Gene Symbol: Dgki | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081206.1 | NP_001074675.1 | 17, 18 | 1765 | ||
| GenBank mRNA | AK081360.1 | - | 17, 18 | 1766 | ||
| AK086550.1 | - | 4, 5 | 357 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dgki| Gene Symbol: Dgki | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081206.1 | NP_001074675.1 | 17, 18 | 1765 | ||
| GenBank mRNA | AK081360.1 | - | 17, 18 | 1766 | ||
| AK086550.1 | - | 4, 5 | 357 | |||