siRNA Details
| Gene Symbol: Fndc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165965.1 | NP_001159437.1 | 9 | 1955 | ||
| NM_177091.5 | NP_796065.2 | 10 | 2213 | |||
| GenBank mRNA | AK053989.1 | - | 10 | 2076 | ||
| AK054241.1 | - | 10 | 2213 | |||
| BC111898.1 | - | 9 | 1762 | 40061374 | ||
| BC113188.1 | - | 10 | 2019 | |||

Fndc7
Genomic Map