siRNA Details
| Gene Symbol: Ddx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003919.1 | NP_001003919.1 | 11 | 1353 | ||
| GenBank mRNA | AK049419.1 | - | 11 | 1359 | ||
| AK076357.1 | - | 12 | 1383 | |||
| AK152868.1 | - | 11 | 1341 | |||
| BC079656.1 | - | 11 | 1353 | 6854039 | ||

Ddx11
Genomic Map