siRNA Details
| Gene Symbol: Senp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177103.4 | NP_796077.2 | 2 | 993 | ||
| GenBank mRNA | AK028177.1 | - | 2 | 994 | ||
| AK042087.1 | - | 2 | 885 | |||
| AK043171.1 | - | 2 | 878 | |||
| AK136120.1 | - | 1 | 586 | |||
| AK158338.1 | - | 2 | 858 | |||
| AY522918.1 | - | 2 | 857 | |||
| BC067014.1 | - | 2 | 760 | 6812169 | ||
| BC080830.1 | - | 2 | 877 | 30433573 | ||

Senp5
Genomic Map