siRNA Details
| Gene Symbol: Ccdc66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177111.3 | NP_796085.3 | 6, 7 | 846 | ||
| GenBank mRNA | AK145297.1 | - | 5, 6 | 859 | ||
| AK158666.1 | - | 4, 5 | 364 | |||
| BC070471.1 | - | 6, 7 | 846 | 30626499 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc66| Gene Symbol: Ccdc66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177111.3 | NP_796085.3 | 6, 7 | 846 | ||
| GenBank mRNA | AK145297.1 | - | 5, 6 | 859 | ||
| AK158666.1 | - | 4, 5 | 364 | |||
| BC070471.1 | - | 6, 7 | 846 | 30626499 | ||