siRNA Details
| Gene Symbol: March3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177115.2 | NP_796089.1 | 2 | 419 | ||
| GenBank mRNA | AK041088.1 | - | 2 | 419 | ||
| BC107232.1 | - | 2 | 203 | 40052073 | ||
| BC107233.1 | - | 2 | 203 | 40052074 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

March3| Gene Symbol: March3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177115.2 | NP_796089.1 | 2 | 419 | ||
| GenBank mRNA | AK041088.1 | - | 2 | 419 | ||
| BC107232.1 | - | 2 | 203 | 40052073 | ||
| BC107233.1 | - | 2 | 203 | 40052074 | ||