siRNA Details
| Gene Symbol: Spef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048961.1 | - | 11 | 1688 | ||
| BC132098.1 | - | 11 | 1625 | 40130375 | ||
| BC132288.1 | - | 11 | 1625 | 40130565 | ||
| HQ856050.1 | - | 12 | 1759 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spef2| Gene Symbol: Spef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048961.1 | - | 11 | 1688 | ||
| BC132098.1 | - | 11 | 1625 | 40130375 | ||
| BC132288.1 | - | 11 | 1625 | 40130565 | ||
| HQ856050.1 | - | 12 | 1759 | |||