siRNA Details
| Gene Symbol: Spef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041992.1 | - | 7 | 952 | ||
| AK048961.1 | - | 7 | 1043 | |||
| BC132098.1 | - | 7 | 980 | 40130375 | ||
| BC132288.1 | - | 7 | 980 | 40130565 | ||
| HQ856050.1 | - | 8 | 1114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spef2| Gene Symbol: Spef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041992.1 | - | 7 | 952 | ||
| AK048961.1 | - | 7 | 1043 | |||
| BC132098.1 | - | 7 | 980 | 40130375 | ||
| BC132288.1 | - | 7 | 980 | 40130565 | ||
| HQ856050.1 | - | 8 | 1114 | |||