siRNA Details
| Gene Symbol: Fryl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028194.2 | NP_082470.2 | 17 | 1945 | ||
| GenBank mRNA | AK036035.1 | - | 15 | 1663 | ||
| AK164590.1 | - | 17 | 1947 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fryl| Gene Symbol: Fryl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028194.2 | NP_082470.2 | 17 | 1945 | ||
| GenBank mRNA | AK036035.1 | - | 15 | 1663 | ||
| AK164590.1 | - | 17 | 1947 | |||