siRNA Details
Gene Symbol: Fryl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028194.2 | NP_082470.2 | 16 | 1893 | ||
GenBank mRNA | AK036035.1 | - | 14 | 1611 | ||
AK164590.1 | - | 16 | 1895 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fryl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028194.2 | NP_082470.2 | 16 | 1893 | ||
GenBank mRNA | AK036035.1 | - | 14 | 1611 | ||
AK164590.1 | - | 16 | 1895 |