siRNA Details
| Gene Symbol: Pde4dip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051209.1 | - | 1, 2 | 292 | ||
| AK083867.1 | - | 1, 2 | 289 | |||
| AK141917.1 | - | 1, 2 | 340 | |||
| CJ125705.1 | - | 1, 2 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde4dip| Gene Symbol: Pde4dip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051209.1 | - | 1, 2 | 292 | ||
| AK083867.1 | - | 1, 2 | 289 | |||
| AK141917.1 | - | 1, 2 | 340 | |||
| CJ125705.1 | - | 1, 2 | 406 | |||