siRNA Details
| Gene Symbol: Bcorl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178782.4 | NP_848897.3 | 8 | 4288 | ||
| GenBank mRNA | AK031119.1 | - | 4 | 781 | ||
| AK054259.1 | - | 4 | 822 | |||
| AK081004.1 | - | 4 | 2856 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcorl1| Gene Symbol: Bcorl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178782.4 | NP_848897.3 | 8 | 4288 | ||
| GenBank mRNA | AK031119.1 | - | 4 | 781 | ||
| AK054259.1 | - | 4 | 822 | |||
| AK081004.1 | - | 4 | 2856 | |||