siRNA Details
| Gene Symbol: Lrig3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177152.5 | NP_796126.4 | 14, 15 | 2340 | ||
| GenBank mRNA | AK018522.1 | - | 8, 9 | 1153 | ||
| AK129481.1 | - | 10, 11 | 1289 | |||
| AY505341.1 | - | 14, 15 | 2283 | |||
| BC065142.1 | - | 14, 15 | 2306 | 30610561 | ||

Lrig3
Genomic Map