siRNA Details
| Gene Symbol: Lrig3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177152.5 | NP_796126.4 | 17 | 3001 | ||
| GenBank mRNA | AK018522.1 | - | 11 | 1814 | ||
| AK020260.1 | - | 1 | 1429 | |||
| AK078783.1 | - | 3 | 595 | |||
| AK135388.1 | - | 1 | 634 | |||
| AY505341.1 | - | 17 | 2944 | |||
| BC065142.1 | - | 17 | 2967 | 30610561 | ||

Lrig3
Genomic Map