siRNA Details
| Gene Symbol: Rinl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177158.5 | NP_796132.2 | 2, 3 | 117 | ||
| GenBank mRNA | AK030993.1 | - | 2, 3 | 110 | ||
| BC044761.1 | - | 2, 3 | 153 | |||
| BC046530.1 | - | 2, 3 | 153 | 5329601 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rinl| Gene Symbol: Rinl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177158.5 | NP_796132.2 | 2, 3 | 117 | ||
| GenBank mRNA | AK030993.1 | - | 2, 3 | 110 | ||
| BC044761.1 | - | 2, 3 | 153 | |||
| BC046530.1 | - | 2, 3 | 153 | 5329601 | ||