siRNA Details
Gene Symbol: Tmprss11g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177162.4 | NP_796136.2 | 9 | 1075 | ||
GenBank mRNA | AK036981.1 | - | 9 | 1076 | ||
AK037029.1 | - | 5 | 573 | |||
BC118611.1 | - | 9 | 1053 | 40107184 | ||
BC118660.1 | - | 9 | 1052 | 40107187 |
