siRNA Details
| Gene Symbol: Heatr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081179.1 | NP_001074648.1 | 5 | 620 | ||
| GenBank mRNA | AK053037.1 | - | 5 | 604 | ||
| AK079542.1 | - | 5 | 565 | |||
| AK146166.1 | - | 5 | 601 | |||
| AK158478.1 | - | 5 | 605 | |||
| AK220390.1 | - | 2 | 153 | |||
| BC086652.1 | - | 4 | 556 | 6856641 | ||

Heatr5b
Genomic Map