siRNA Details
| Gene Symbol: A830018L16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160369.2 | NP_001153841.1 | 13 | 1792 | ||
| NM_177173.6 | NP_796147.2 | 13 | 1783 | |||
| GenBank mRNA | AK034391.1 | - | 13 | 1779 | ||
| BC137903.1 | - | 13 | 1388 | 8860922 | ||
| BC145022.1 | - | 13 | 1388 | 9053570 | ||
| BC145023.1 | - | 12 | 1340 | 9053571 | ||

A830018L16Rik
Genomic Map