siRNA Details
| Gene Symbol: Lmod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081157.1 | NP_001074626.1 | 2 | 1398 | ||
| GenBank mRNA | AK030676.1 | - | 2 | 1380 | ||
| BC127157.1 | - | 2 | 1326 | 40091077 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmod3| Gene Symbol: Lmod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081157.1 | NP_001074626.1 | 2 | 1398 | ||
| GenBank mRNA | AK030676.1 | - | 2 | 1380 | ||
| BC127157.1 | - | 2 | 1326 | 40091077 | ||