siRNA Details
| Gene Symbol: Slc35e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177186.4 | NP_796160.1 | 4 | 981 | ||
| GenBank mRNA | AK048716.1 | - | 4 | 850 | ||
| AK080215.1 | - | 4 | 1000 | |||
| AK145065.1 | - | 5 | 904 | |||
| AK157354.1 | - | 4 | 877 | |||
| BC058728.1 | - | 4 | 981 | 6407259 | ||
| Gene Symbol: Gm10563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK155949.1 | - | 1 | 3370 | ||

Slc35e2
Genomic Map