siRNA Details
| Gene Symbol: Snrnp200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177214.4 | NP_796188.2 | 35 | 5072 | ||
| GenBank mRNA | AK149291.1 | - | 6 | 595 | ||
| AK173025.1 | - | 30 | 4301 | |||
| BC011390.1 | - | 8 | 1262 | |||
| BC023685.1 | - | 8 | 1186 | |||
| BC030856.1 | - | 7 | 1013 | |||
| BC058608.1 | - | 4 | 555 | |||
| BC063261.1 | - | 35 | 5072 | 6408607 | ||

Snrnp200
Genomic Map