siRNA Details
| Gene Symbol: D5Ertd579e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081232.3 | NP_001074701.3 | 6 | 3731 | ||
| GenBank mRNA | AK122224.1 | - | 5 | 3242 | ||
| AK139370.1 | - | 6 | 3648 | |||
| BC094673.1 | - | 1 | 2264 | |||
| BC151085.1 | - | 5 | 3476 | 9087998 | ||

D5Ertd579e
Genomic Map