siRNA Details
| Gene Symbol: Casc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177222.4 | NP_796196.3 | 9 | 1018 | ||
| GenBank mRNA | AK034148.1 | - | 8 | 866 | ||
| AK132912.1 | - | 9 | 1242 | |||
| AY423542.1 | - | 9 | 949 | |||
| AY485607.1 | - | 9 | 1018 | |||
| AY485608.1 | - | 9 | 1018 | |||
| BC072568.1 | - | 7 | 860 | |||
| BC120743.1 | - | 9 | 922 | 40129666 | ||
| BC125423.1 | - | 9 | 922 | 40129938 | ||
| BC144801.1 | - | 9 | 922 | |||

Casc1
Genomic Map