siRNA Details
| Gene Symbol: Tmem117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178789.4 | NP_848904.1 | 6 | 891 | ||
| GenBank mRNA | AK047437.1 | - | 6 | 892 | ||
| AK048597.1 | - | 6 | 851 | |||
| AK052935.1 | - | 4 | 483 | |||
| BC051178.1 | - | 6 | 826 | 6314268 | ||
| BC054070.1 | - | 6 | 826 | |||

Tmem117
Genomic Map