siRNA Details
| Gene Symbol: Lingo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177250.2 | NP_796224.1 | 2 | 588 | ||
| GenBank mRNA | AK040952.1 | - | 2 | 588 | ||
| AK042286.1 | - | 1289 | ||||
| BC117866.1 | - | 2 | 498 | 40095465 | ||
| BC117867.1 | - | 2 | 498 | 40095468 | ||

Lingo4
Genomic Map