siRNA Details
| Gene Symbol: L3mbtl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177278.5 | NP_796252.2 | 5, 6 | 808 | ||
| GenBank mRNA | AK039470.1 | - | 5, 6 | 779 | ||
| AK053068.1 | - | 5, 6 | 809 | |||
| BC147118.1 | - | 4, 5 | 571 | |||
| BC147119.1 | - | 4, 5 | 571 | |||

L3mbtl4
Genomic Map