siRNA Details
| Gene Symbol: L3mbtl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177278.5 | NP_796252.2 | 4, 5 | 698 | ||
| GenBank mRNA | AK039470.1 | - | 4, 5 | 669 | ||
| AK053068.1 | - | 4, 5 | 699 | |||
| BC147118.1 | - | 3, 4 | 461 | |||
| BC147119.1 | - | 3, 4 | 461 | |||

L3mbtl4
Genomic Map