siRNA Details
| Gene Symbol: Krt26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033397.5 | NP_001028569.2 | 7 | 1254 | ||
| GenBank mRNA | AK028591.1 | - | 7 | 1254 | ||
| AK028682.1 | - | 6 | 2387 | |||
| AK132445.1 | - | 7 | 1258 | |||
| AK162718.1 | - | 7 | 1256 | |||
| BC116672.1 | - | 7 | 1243 | 40107596 | ||

Krt26
Genomic Map