siRNA Details
Gene Symbol: Krt26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033397.5 | NP_001028569.2 | 8 | 1328 | ||
GenBank mRNA | AK028591.1 | - | 8 | 1328 | ||
AK028682.1 | - | 7 | 2461 | |||
AK132445.1 | - | 8 | 1332 | |||
AK162718.1 | - | 8 | 1330 | |||
BC116672.1 | - | 8 | 1317 | 40107596 |
