siRNA Details
Gene Symbol: Tnpo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177296.4 | NP_796270.2 | 2 | 291 | ||
GenBank mRNA | AK045143.1 | - | 2 | 319 | ||
AK049446.1 | - | 2 | 505 | |||
AK145723.1 | - | 2 | 308 | |||
BC064646.1 | - | 2 | 291 | 6313452 | ||
BC075678.1 | - | 2 | 504 | 30619729 |
