siRNA Details
| Gene Symbol: Tnpo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177296.4 | NP_796270.2 | 18, 19 | 2385 | ||
| GenBank mRNA | AK077714.1 | - | 2, 3 | 215 | ||
| AK220184.1 | - | 11, 12 | 1288 | |||
| BC023273.1 | - | 6, 7 | 570 | |||
| BC052756.1 | - | 7, 8 | 706 | |||
| BC064646.1 | - | 18, 19 | 2385 | 6313452 | ||
| BC075678.1 | - | 18, 19 | 2598 | 30619729 | ||

Tnpo3
Genomic Map