siRNA Details
| Gene Symbol: Cyp4f39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177307.3 | NP_796281.1 | 10 | 1228 | ||
| GenBank mRNA | AK028950.1 | - | 10 | 1229 | ||
| AK037192.1 | - | 10 | 1229 | |||
| BC145755.1 | - | 10 | 1225 | 40131058 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp4f39| Gene Symbol: Cyp4f39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177307.3 | NP_796281.1 | 10 | 1228 | ||
| GenBank mRNA | AK028950.1 | - | 10 | 1229 | ||
| AK037192.1 | - | 10 | 1229 | |||
| BC145755.1 | - | 10 | 1225 | 40131058 | ||