siRNA Details
| Gene Symbol: 1700006J14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005678.1 | - | 3 | 606 | ||
| AK011105.1 | - | 4 | 646 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_177313.1 | - | 606 | Chr. - - [-] on Build | ||

1700006J14Rik
Genomic Map