siRNA Details
| Gene Symbol: Upf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081132.1 | NP_001074601.1 | 21 | 3876 | ||
| GenBank mRNA | BC039278.1 | - | 10 | 1532 | ||
| BC138358.1 | - | 21 | 3839 | 8861378 | ||
| BC138359.1 | - | 21 | 3839 | 8861379 | ||

Upf2
Genomic Map