siRNA Details
| Gene Symbol: Tmem26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177794.3 | NP_808462.2 | 3, 4 | 548 | ||
| GenBank mRNA | AK028283.1 | - | 3, 4 | 549 | ||
| AK084351.1 | - | 3, 4 | 526 | |||
| AK143866.1 | - | 3, 4 | 526 | |||
| BC117502.1 | - | 4, 5 | 552 | 40106131 | ||
| BC117503.1 | - | 3, 4 | 470 | 40106128 | ||

Tmem26
Genomic Map