siRNA Details
| Gene Symbol: Pitpnm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024927.2 | NP_001020098.1 | 9 | 1284 | ||
| NM_001081641.1 | NP_001075110.1 | 8 | 1236 | |||
| GenBank mRNA | AK039594.1 | - | 8 | 1477 | ||
| AK147444.1 | - | 9 | 1241 | |||
| AK147630.1 | - | 8 | 1069 | |||
| BC112411.1 | - | 8 | 1659 | 40061037 | ||

Pitpnm3
Genomic Map